Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARL16
Entrez Gene ID
101116745
Gene Name
ADP ribosylation factor like GTPase 16
Gene Aliases
-
Chromosome Location
Chr.11: 11976797 - 11979579 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027974270.1 XP_027830071.1 2 - 3 159 100

View Full Product Details