Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C15H11orf94
Entrez Gene ID
101117191
Gene Name
chromosome 15 C11orf94 homolog
Gene Aliases
-
Chromosome Location
Chr.15: 82189223 - 82189859 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027979657.1 XP_027835458.1 2 - 3 187 104

View Full Product Details