Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
METTL9
Entrez Gene ID
101115339
Gene Name
methyltransferase like 9
Gene Aliases
-
Chromosome Location
Chr.24: 20784004 - 20836700 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015104122.2 XP_014959608.1 4 - 5 1108 103
XM_015104124.2 XP_014959610.2 4 - 5 1062 103

View Full Product Details