Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TAS2R8
Entrez Gene ID
101115357
Gene Name
taste 2 receptor member 8
Gene Aliases
-
Chromosome Location
Chr.3: 219068083 - 219069011 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012176118.3 XP_012031508.3 1 - 1 683 61

View Full Product Details