The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_012102275.3 | XP_011957665.2 | 4 - 5 | 413 | 84 |
XM_015103087.2 | XP_014958573.1 | 4 - 5 | 487 | 84 |