Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC97
Entrez Gene ID
101102506
Gene Name
coiled-coil domain containing 97
Gene Aliases
-
Chromosome Location
Chr.14: 53710564 - 53721085 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004015277.4 XP_004015326.3 3 - 4 937 58

View Full Product Details