Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AK4
Entrez Gene ID
101103031
Gene Name
adenylate kinase 4
Gene Aliases
-
Chromosome Location
Chr.1: 42691654 - 42783058 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027970883.1 XP_027826684.1 4 - 5 724 67

View Full Product Details