Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
IGSF8
Entrez Gene ID
101102283
Gene Name
immunoglobulin superfamily member 8
Gene Aliases
-
Chromosome Location
Chr.1: 117966985 - 117974018 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027976987.1 XP_027832788.1 1 - 2 230 64

View Full Product Details