Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RWDD2B
Entrez Gene ID
101109662
Gene Name
RWD domain containing 2B
Gene Aliases
-
Chromosome Location
Chr.1: 135403660 - 135417264 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004002791.4 XP_004002840.1 4 - 5 1279 63

View Full Product Details