Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IL20
Entrez Gene ID
101112916
Gene Name
interleukin 20
Gene Aliases
-
Chromosome Location
Chr.12: 4266212 - 4269156 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004013961.4 XP_004014010.3 2 - 3 220 98

View Full Product Details