Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FUS
Entrez Gene ID
101104799
Gene Name
FUS RNA binding protein
Gene Aliases
-
Chromosome Location
Chr.24: 28033439 - 28043644 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012104405.3 XP_011959795.2 3 - 4 270 149

View Full Product Details