Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRMT5
Entrez Gene ID
101105080
Gene Name
tRNA methyltransferase 5
Gene Aliases
-
Chromosome Location
Chr.7: 76041716 - 76052112 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004010700.3 XP_004010749.2 3 - 4 1065 112

View Full Product Details