Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MTSS2
Entrez Gene ID
101114381
Gene Name
MTSS I-BAR domain containing 2
Gene Aliases
MTSS1L
Chromosome Location
Chr.14: 1020653 - 1040440 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027977613.1 XP_027833414.1 12 - 13 1185 100

View Full Product Details