Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GID8
Entrez Gene ID
101111114
Gene Name
GID complex subunit 8 homolog
Gene Aliases
-
Chromosome Location
Chr.13: 56824324 - 56832394 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004014414.4 XP_004014463.1 2 - 3 384 75
XM_027976962.1 XP_027832763.1 2 - 3 333 75
XM_027976963.1 XP_027832764.1 2 - 3 511 75

View Full Product Details