Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ELAC1
Entrez Gene ID
101118238
Gene Name
elaC ribonuclease Z 1
Gene Aliases
-
Chromosome Location
Chr.23: 55906333 - 55926238 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004020546.3 XP_004020595.1 2 - 3 232 103
XM_004020547.3 XP_004020596.1 2 - 3 896 103

View Full Product Details