Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
REXO4
Entrez Gene ID
101112473
Gene Name
REX4 homolog, 3'-5' exonuclease
Gene Aliases
-
Chromosome Location
Chr.3: 2677731 - 2685753 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027966150.1 XP_027821951.1 2 - 3 608 96

View Full Product Details