Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RSPH6A
Entrez Gene ID
101118893
Gene Name
radial spoke head 6 homolog A
Gene Aliases
-
Chromosome Location
Chr.14: 56822296 - 56846480 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015100604.2 XP_014956090.2 2 - 3 983 104
XM_027978495.1 XP_027834296.1 2 - 3 983 104

View Full Product Details