Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CHCHD1
Entrez Gene ID
101120800
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 1
Gene Aliases
-
Chromosome Location
Chr.25: 30764240 - 30765560 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004021478.3 XP_004021527.1 1 - 2 172 117

View Full Product Details