Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ANKLE1
Entrez Gene ID
101115620
Gene Name
ankyrin repeat and LEM domain containing 1
Gene Aliases
-
Chromosome Location
Chr.5: 5979854 - 5984463 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012177715.3 XP_012033105.2 2 - 3 374 72

View Full Product Details