Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SWI5
Entrez Gene ID
101120905
Gene Name
SWI5 homologous recombination repair protein
Gene Aliases
-
Chromosome Location
Chr.3: 8268799 - 8273811 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012162142.1 XP_012017532.1 1 - 2 141 100

View Full Product Details