Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
P4HTM
Entrez Gene ID
101109046
Gene Name
prolyl 4-hydroxylase, transmembrane
Gene Aliases
-
Chromosome Location
Chr.19: 52963119 - 52976897 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027957663.1 XP_027813464.1 2 - 3 430 94

View Full Product Details