Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OMP
Entrez Gene ID
101113008
Gene Name
olfactory marker protein
Gene Aliases
-
Chromosome Location
Chr.15: 59840795 - 59841286 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012096098.3 XP_011951488.1 1 - 1 216 67

View Full Product Details