Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RCVRN
Entrez Gene ID
101111206
Gene Name
recoverin
Gene Aliases
-
Chromosome Location
Chr.11: 34395796 - 34403641 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015098653.2 XP_014954139.1 2 - 3 488 81

View Full Product Details