Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCND1
Entrez Gene ID
101106661
Gene Name
potassium voltage-gated channel subfamily D member 1
Gene Aliases
-
Chromosome Location
Chr.X: 58447573 - 58456263 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004022096.4 XP_004022145.2 5 - 6 3474 100

View Full Product Details