Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TADA3
Entrez Gene ID
101103513
Gene Name
transcriptional adaptor 3
Gene Aliases
-
Chromosome Location
Chr.19: 17922990 - 17933421 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027958097.1 XP_027813898.1 8 - 9 1065 79
XM_027958098.1 XP_027813899.1 8 - 9 1062 79
XM_027958099.1 XP_027813900.1 8 - 9 1062 79
XM_027958102.1 XP_027813903.1 8 - 9 1068 79

View Full Product Details