Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WIPF3
Entrez Gene ID
101111451
Gene Name
WAS/WASL interacting protein family member 3
Gene Aliases
-
Chromosome Location
Chr.4: 73201783 - 73228462 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027968646.1 XP_027824447.1 6 - 7 2032 89

View Full Product Details