Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRRC66
Entrez Gene ID
101104319
Gene Name
leucine rich repeat containing 66
Gene Aliases
-
Chromosome Location
Chr.6: 74426455 - 74450600 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027970994.1 XP_027826795.1 3 - 4 853 85

View Full Product Details