Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKRD11
Entrez Gene ID
101110169
Gene Name
ankyrin repeat domain 11
Gene Aliases
-
Chromosome Location
Chr.14: 15112950 - 15230198 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027977755.1 XP_027833556.1 6 - 7 807 59
XM_027977756.1 XP_027833557.1 6 - 7 810 59
XM_027977757.1 XP_027833558.1 6 - 7 807 59
XM_027977758.1 XP_027833559.1 4 - 5 593 59

View Full Product Details