Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FUNDC1
Entrez Gene ID
101107165
Gene Name
FUN14 domain containing 1
Gene Aliases
-
Chromosome Location
Chr.X: 45265860 - 45278252 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004022019.4 XP_004022068.1 4 - 5 552 83

View Full Product Details