Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZBTB8A
Entrez Gene ID
101112885
Gene Name
zinc finger and BTB domain containing 8A
Gene Aliases
-
Chromosome Location
Chr.2: 250636753 - 250697478 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027965343.1 XP_027821144.1 4 - 5 1432 77

View Full Product Details