Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ASPSCR1
Entrez Gene ID
101107539
Gene Name
ASPSCR1 tether for SLC2A4, UBX domain containing
Gene Aliases
-
Chromosome Location
Chr.11: 12189893 - 12218236 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015098906.2 XP_014954392.2 3 - 4 326 116
XM_027974287.1 XP_027830088.1 3 - 4 534 116
XM_027974288.1 XP_027830089.1 3 - 4 296 116
XM_027974289.1 XP_027830090.1 2 - 3 249 116

View Full Product Details