Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LHX1
Entrez Gene ID
101109722
Gene Name
LIM homeobox 1
Gene Aliases
-
Chromosome Location
Chr.11: 50624908 - 50630707 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027975315.1 XP_027831116.1 1 - 2 688 104
XM_027975316.1 XP_027831117.1 2 - 3 1220 104

View Full Product Details