Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MARCHF4
Entrez Gene ID
101107500
Gene Name
membrane associated ring-CH-type finger 4
Gene Aliases
MARCH4
Chromosome Location
Chr.2: 233055311 - 233175698 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027964854.1 XP_027820655.1 6 - 7 991 81

View Full Product Details