Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CDK3
Entrez Gene ID
101107113
Gene Name
cyclin dependent kinase 3
Gene Aliases
-
Chromosome Location
Chr.11: 7617491 - 7621821 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027974103.1 XP_027829904.1 5 - 6 1259 85

View Full Product Details