Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FOXO6
Entrez Gene ID
101123434
Gene Name
forkhead box O6
Gene Aliases
-
Chromosome Location
Chr.1: 16479947 - 16500287 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027968478.1 XP_027824279.1 1 - 2 416 114

View Full Product Details