Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C4H7orf25
Entrez Gene ID
101120235
Gene Name
chromosome 4 C7orf25 homolog
Gene Aliases
-
Chromosome Location
Chr.4: 85717493 - 85720510 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027968749.1 XP_027824550.1 2 - 2 674 108

View Full Product Details