Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GSPT1
Entrez Gene ID
100008618
Gene Name
G1 to S phase transition 1
Gene Aliases
ELF
Chromosome Location
Chr.6: 5287173 - 5315685 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_002711756.3 XP_002711802.1 11 - 12 1649 94
XM_008257735.2 XP_008255957.1 11 - 12 1646 94
GenBank mRNA AB035256.1 - 11 - 12 1281 94

View Full Product Details