Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRKG2
Entrez Gene ID
100008781
Gene Name
protein kinase cGMP-dependent 2
Gene Aliases
CGK-II
Chromosome Location
Chr.15: 69077296 - 69185870 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_002717087.3 XP_002717133.1 2 - 2 351 89
GenBank mRNA AF274319.1 - 1 - 1 146 89

View Full Product Details