Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FOXJ1
Entrez Gene ID
100008873
Gene Name
forkhead box J1
Gene Aliases
-
Chromosome Location
Chr.Unknown: 537975 - 542830 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_002722923.3 XP_002722969.2 - 895 93
GenBank mRNA AM056025.1 - - 70 93

View Full Product Details