The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198981.1 | NP_001185910.1 | 1 - 1 | 82 | |
XM_008258577.2 | XP_008256799.1 | 1 - 1 | 82 | ||
GenBank mRNA | EF428024.1 | - | 1 - 1 | 52 | 82 |