Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ITIH4
Entrez Gene ID
100008625
Gene Name
inter-alpha-trypsin inhibitor heavy chain 4
Gene Aliases
ITI-HC4
Chromosome Location
Chr.9: 19842961 - 19857724 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082009.1 NP_001075478.1 12 - 13 1686 73
XM_017343628.1 XP_017199117.1 12 - 13 1701 73
XM_017343629.1 XP_017199118.1 12 - 13 1701 73
XM_017343630.1 XP_017199119.1 12 - 13 1701 73
XM_017343631.1 XP_017199120.1 12 - 13 1702 73
XM_017343632.1 XP_017199121.1 12 - 13 1702 73
GenBank mRNA AB050594.1 - 12 - 13 1686 73

View Full Product Details