Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HNRNPC
Entrez Gene ID
100008686
Gene Name
heterogeneous nuclear ribonucleoprotein C
Gene Aliases
HNRPC
Chromosome Location
Chr.17: 41457631 - 41512452 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082038.1 NP_001075507.1 7 - 8 809 138
XM_008268785.2 XP_008267007.1 8 - 9 928 138
XM_008268786.2 XP_008267008.1 7 - 8 900 138
XM_008268787.2 XP_008267009.1 6 - 7 854 138
XM_008268789.2 XP_008267011.1 8 - 9 889 138
XM_017347898.1 XP_017203387.1 8 - 9 928 138
GenBank mRNA AF061582.1 - 7 - 8 809 138

Gene Symbol
LOC100344160
Entrez Gene ID
100344160
Gene Name
heterogeneous nuclear ribonucleoprotein C-like
Gene Aliases
-
Chromosome Location
Chr.12: 77922057 - 77923477 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_002714588.3 XP_002714634.1 2 - 2 809 138
XM_008263221.2 XP_008261443.1 2 - 2 846 138
XM_017345284.1 XP_017200773.1 2 - 2 767 138

Gene Symbol
LOC100354921
Entrez Gene ID
100354921
Gene Name
heterogeneous nuclear ribonucleoprotein C
Gene Aliases
-
Chromosome Location
Chr.Unknown: 4627 - 6054 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008253032.2 XP_008251254.1 7 - 8 849 138
XM_008253033.2 XP_008251255.1 8 - 9 832 138

View Full Product Details