Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VCAM1
Entrez Gene ID
100008901
Gene Name
vascular cell adhesion molecule 1
Gene Aliases
VCAM-1
Chromosome Location
Chr.13: 63440580 - 63462573 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082152.1 NP_001075621.1 2 - 3 447 98
XM_008263958.2 XP_008262180.1 2 - 3 673 98
GenBank mRNA AB128156.1 - 1 - 2 166 98
AY212510.1 - 2 - 3 447 98

View Full Product Details