Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACO1
Entrez Gene ID
100009154
Gene Name
aconitase 1
Gene Aliases
FRP
Chromosome Location
Chr.1: 21028534 - 21086931 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082315.1 NP_001075784.1 14 - 15 1785 79
XM_008248341.2 XP_008246563.1 14 - 15 1847 79
XM_008253164.2 XP_008251386.1 13 - 14 1747 79
GenBank mRNA M95815.1 - 14 - 15 1785 79

View Full Product Details