Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CA4
Entrez Gene ID
100009226
Gene Name
carbonic anhydrase 4
Gene Aliases
-
Chromosome Location
Chr.19: 26488730 - 26492286 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082372.1 NP_001075841.1 3 - 4 452 73
GenBank mRNA L48928.1 - 3 - 4 411 73
U58871.1 - 3 - 4 452 73

View Full Product Details