Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PTGS1
Entrez Gene ID
100009407
Gene Name
prostaglandin-endoperoxide synthase 1
Gene Aliases
COX-1
Chromosome Location
Chr.Unknown: 4844436 - 4866579 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082681.1 NP_001076150.1 - 542 102
GenBank mRNA AF026008.1 - - 542 102

View Full Product Details