Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PGA5
Entrez Gene ID
100009429
Gene Name
pepsinogen 5, group I (pepsinogen A)
Gene Aliases
-
Chromosome Location
Chr.Unknown: 3893731 - 3904163 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082696.1 NP_001076165.1 - 927 80
GenBank mRNA M59238.1 - - 927 80

View Full Product Details