The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001082787.2 | NP_001076256.1 | 2 - 3 | 268 | 147 |
XM_008255647.2 | XP_008253869.1 | 3 - 4 | 463 | 147 | |
GenBank mRNA | AY101395.1 | - | 2 - 3 | 270 | 147 |