Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPP1CB
Entrez Gene ID
100009587
Gene Name
protein phosphatase 1 catalytic subunit beta
Gene Aliases
PP1-BETA
Chromosome Location
Chr.2: 157289451 - 157337162 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101722.1 NP_001095192.1 2 - 3 183 108
GenBank mRNA X61639.1 - - 183 108

View Full Product Details