Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
JPH2
Entrez Gene ID
100008613
Gene Name
junctophilin 2
Gene Aliases
JP-2, jp
Chromosome Location
Chr.Unknown: 3430614 - 3458106 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081998.1 NP_001075467.1 - 1049 89
GenBank mRNA AB024444.1 - - 1049 89

View Full Product Details